水, UltraPure™ DNase-, RNase-free, distilled
供应商: INVITROGEN
Synonyms:
Hydrogen oxide
10977015
10977023
INVT10977015EA
0
CNY
INVT10977015
INVT10977023
水, UltraPure™ DNase-, RNase-free, distilled
水
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
分子式:
H₂O 沸点: 100 °C (1013 hPa) 溶点: 0 °C 密度: 1 g/cm³ (4 °C) 存储条件: Ambient |
MDL编号:
MFCD00011332 CAS号: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...