占吨醇 97+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 氧杂蒽醇, 9-羟基呫吨

271520050 271520250
ACRO271520050EA 0 CNY
ACRO271520050 ACRO271520250
占吨醇 97+%
占吨醇
分子式: C₁₃H₁₀O₂
沸点: 338 °C (1013 hPa)
溶点: 124…127 °C
存储条件: Refrigerator
MDL编号: MFCD00005057
CAS号: 90-46-0
EINECS: 201-996-1

现在订购

产品技术规格

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 121 to 126 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR