9,10-菲醌 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 9,10-菲二酮

168790250 168791000
ACRO168790250EA 0 CNY
ACRO168790250 ACRO168791000
9,10-菲醌 95%
9,10-菲醌
分子式: C₁₄H₈O₂
分子量: 208.22 g/mol
沸点: 360
溶点: 201…211 °C
闪点: 245 °C
存储条件: Ambient
MDL编号: MFCD00001163
CAS号: 84-11-7
EINECS: 201-515-5
Merck Index: 12,07355

现在订购

产品技术规格

Appearance Orange-brown Powder
Infrared spectrum Conforms
Melting point 203 to 211 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR