顺-环辛烯 95% stabilised

供应商: Thermo Fisher Scientific
Danger

Synonyms: (Z)-Cyclooctene

154860010 154862500
ACRO154860010EA 0 CNY
ACRO154860010 ACRO154862500
顺-环辛烯 95% stabilised
顺-环辛烯
分子式: C₈H₁₄
分子量: 110.2 g/mol
沸点: 145…146 °C (760 mmHg)
溶点: –16 °C
密度: 0.84 g/cm³
闪点: 25 °C
存储条件: Ambient
MDL编号: MFCD00001753
CAS号: 931-87-3
EINECS: 213-243-4
UN号: 3295
ADR: 3 III

现在订购

产品技术规格

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4710 (20°C, 589 nm)
GC ≥94 %
Stabilizer 100 to 200 ppm irganox 1076 FD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR