2,2'-联苯酚 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2,2′-联苯二酚, 2,2′-联苯酚, 2,2′-二羟基联苯

151991000 151995000
ACRO151991000EA 0 CNY
ACRO151991000 ACRO151995000
2,2'-联苯酚 99%
2,2'-联苯酚
分子式: C₁₂H₁₀O₂
分子量: 186.21 g/mol
沸点: 315
溶点: 108…111 °C
密度: 0.83 g/cm³
闪点: 154 °C
MDL编号: MFCD00002210
CAS号: 1806-29-7
EINECS: 217-303-0

现在订购

产品技术规格

Appearance Beige to light grey Powder
Infrared spectrum Conforms
Melting point 108 to 111 °C
Loss on drying ≤0.5 %
GC ≥98.5 %
Iron (Fe) ≤15 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR