2-苯基苯酚 99+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2-羟基联苯

130760050 130765000
ACRO130760050EA 0 CNY
ACRO130760050 ACRO130765000
2-苯基苯酚 99+%
2-苯基苯酚
分子式: C₁₂H₁₀O
分子量: 170.21 g/mol
沸点: 280…286 °C (1013 hPa)
溶点: >56 °C
密度: 1.21 g/cm³
闪点: 123 °C
存储条件: Ambient
MDL编号: MFCD00002208
CAS号: 90-43-7
EINECS: 201-993-5
UN号: 3077
ADR: 9 III
Merck Index: 12,07458

现在订购

产品技术规格

Appearance White to off-white Crystalline flakes
Infrared spectrum Conforms
Melting point 56 to 59 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR