1,3-苯二甲醇 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 间二甲苯-α,α′-二醇

188200020 188200100
ACRO188200020EA 0 CNY
ACRO188200020 ACRO188200100
1,3-苯二甲醇 98%
1,3-苯二甲醇
分子式: C₈H₁₀O₂
分子量: 138.17 g/mol
沸点: 154…159 °C (13 mmHg)
溶点: 56…60 °C
MDL编号: MFCD00004649
CAS号: 626-18-6
EINECS: 210-934-2

现在订购

产品技术规格

Appearance White to beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR