对甲基苄胺 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
对甲苄胺
126400250
126401000
126400050
ACRO126400250EA
0
CNY
ACRO126400250
ACRO126401000
ACRO126400050
对甲基苄胺 98%
对甲基苄胺
分子式:
C₈H₁₁N 沸点: 195 °C (1013 hPa) 溶点: 13 °C 密度: 0,95 g/cm³ 闪点: 75 °C 存储条件: Ambient |
MDL编号:
MFCD00008123 CAS号: 104-84-7 EINECS: 203-243-2 UN号: 2735 ADR: 8,III |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5330 to 1.5360 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...