1,3-双(三氟甲基)-5-溴苯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-溴-3,5-二(三氟甲基)苯, 3,5-双(三氟甲基)溴苯
307700100
307700500
ACRO307700100EA
0
CNY
ACRO307700100
ACRO307700500
1,3-双(三氟甲基)-5-溴苯 99%
1,3-双(三氟甲基)-5-溴苯
分子式:
C₈H₃BrF₆ 分子量: 293.01 g/mol 沸点: 154 °C 溶点: –16 °C 密度: 1.71 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00000381 CAS号: 328-70-1 EINECS: 206-334-5 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4260 to 1.4280 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...