α,α,α′,α′-四溴邻二甲苯 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1,2-双(二溴甲基)苯
137971000
137970250
ACRO137971000EA
0
CNY
ACRO137971000
ACRO137970250
α,α,α′,α′-四溴邻二甲苯 97%
α,α,α′,α′-四溴邻二甲苯
分子式:
C₈H₆Br₄ 分子量: 421.75 g/mol 沸点: 346 °C (1013 hPa) 溶点: 114…118 °C 存储条件: Ambient |
MDL编号:
MFCD00000131 CAS号: 13209-15-9 EINECS: 236-176-2 UN号: 3261 ADR: 8 II |
产品技术规格
Appearance | Off-white to beige to light grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 114 to 118 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...