对甲苯磺酸甲酯 97%

供应商: Thermo Fisher Scientific
Danger

Synonyms: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester

170101000 170105000
ACRO170101000EA 0 CNY
ACRO170101000 ACRO170105000
对甲苯磺酸甲酯 97%
对甲苯磺酸甲酯
分子式: C₈H₁₀O₃S
分子量: 186.23 g/mol
沸点: 292 °C (760 mmHg)
溶点: 24…30 °C
闪点: >110 °C
存储条件: Ambient
MDL编号: MFCD00008417
CAS号: 80-48-8
EINECS: 201-283-5
UN号: 3261
ADR: 8 III

现在订购

产品技术规格

Appearance White to yellow Low melting solid or liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5190 (20°C, 589 nm)
Melting point 24 to 30°C
Water ≤0.5 % (K.F.)
GC ≥96.0 %
Acidity ≤0.2 %
Titration excess NaOH 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR