对甲苯磺酸乙酯 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

147230010 147232500
ACRO147230010EA 0 CNY
ACRO147230010 ACRO147232500
对甲苯磺酸乙酯 98%
对甲苯磺酸乙酯
分子式: C₉H₁₂O₃S
分子量: 200.26 g/mol
沸点: 158…162 °C (10 mmHg)
溶点: 32…34 °C
密度: 1.174 g/cm³
闪点: 157 °C
存储条件: Ambient
MDL编号: MFCD00009100
CAS号: 80-40-0
EINECS: 201-276-7
Merck Index: 13,03889

现在订购

产品技术规格

Appearance Clear colorless to light brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5120 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR