2-甲基苯硫酚 97%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 邻甲苯硫酚

259240050 259240250
ACRO259240050EA 0 CNY
ACRO259240050 ACRO259240250
2-甲基苯硫酚 97%
2-甲基苯硫酚
分子式: C₇H₈S
沸点: 194 °C (760 mmHg)
溶点: 14 °C
密度: 1,054 g/cm³
闪点: 63 °C
存储条件: Ambient
MDL编号: MFCD00004838
CAS号: 137-06-4
EINECS: 205-276-8
UN号: 3334
ADR: 9,III
Merck Index: 12,09461

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR