对羟基苯甲醇 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

121030250 121031000
ACRO121030250EA 0 CNY
ACRO121030250 ACRO121031000
对羟基苯甲醇 97%
对羟基苯甲醇
分子式: C₇H₈O₂
分子量: 124.14 g/mol
沸点: 252
溶点: 110…112 °C
存储条件: Ambient
MDL编号: MFCD00004658
CAS号: 623-05-2
EINECS: 210-768-0

现在订购

产品技术规格

Appearance Beige to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
TMS 0.028 to 0.032 v/v%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR