2-羟基苄醇 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
邻羟基苄醇, 水杨醇
173330250
173331000
ACRO173330250EA
0
CNY
ACRO173330250
ACRO173331000
2-羟基苄醇 97%
2-羟基苄醇
分子式:
C₇H₈O₂ 分子量: 124.14 g/mol 沸点: 268 溶点: 83…87 °C 存储条件: Ambient |
MDL编号:
MFCD00004617 CAS号: 90-01-7 EINECS: 201-960-5 Merck Index: 12,08477 |
产品技术规格
Appearance | Light beige Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 83 to 87 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...