2-甲氧基苄醇 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 邻茴香醇

125760100 125760500
ACRO125760100EA 0 CNY
ACRO125760100 ACRO125760500
2-甲氧基苄醇 99%
2-甲氧基苄醇
分子式: C₈H₁₀O₂
分子量: 138.17 g/mol
沸点: 248…250 °C (760 mmHg)
密度: 1.03 g/cm³
闪点: >112 °C
存储条件: Ambient
MDL编号: MFCD00004611
CAS号: 612-16-8
EINECS: 210-296-5

现在订购

产品技术规格

Appearance Clear colorless to Yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5490 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR