4-Nitrobenzyl alcohol 99%
供应商: Thermo Fisher Scientific
|
Warning
|
128520250
128521000
ACRO128520250EA
0
CNY
ACRO128520250
ACRO128521000
4-Nitrobenzyl alcohol 99%
4-Nitrobenzyl alcohol
分子式:
C₇H₇NO₃ 沸点: 185 °C (12 mmHg) 溶点: 92…95 °C 闪点: 180 °C 存储条件: Ambient |
MDL编号:
MFCD00007376 CAS号: 619-73-8 EINECS: 210-611-6 |
产品技术规格
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 92 to 95 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...