2-碘代乙基苯 97% stabilised

供应商: Thermo Fisher Scientific

Synonyms: 2-Phenylethyl iodide, (2-Iodoethyl)benzene

Stabilization: Stabilised with copper

305250050 305250250
ACRO305250050EA 0 CNY
ACRO305250050 ACRO305250250
2-碘代乙基苯 97% stabilised
2-碘代乙基苯
分子式: C₆H₅CH₂CH₂I
沸点: 122…123 °C (13 mmHg)
溶点: –2 °C
密度: 1,6 g/cm³
闪点: >110 °C
MDL编号: MFCD00039408
CAS号: 17376-04-4

现在订购

产品技术规格

Appearance Clear colorless to yellow to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5990 to 1.6030 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR