4-(三氟甲氧基)苄胺 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (4-(Trifluoromethoxy)phenyl)methanamine

297700010 297700050
ACRO297700010EA 0 CNY
ACRO297700010 ACRO297700050
4-(三氟甲氧基)苄胺 97%
4-(三氟甲氧基)苄胺
分子式: C₈H₈F₃NO
分子量: 191.15 g/mol
沸点: 57…60 °C (10 mmHg)
密度: 1.25 g/cm³
闪点: 77 °C
MDL编号: MFCD00061237
CAS号: 93919-56-3
EINECS: 300-040-1

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR