4-(三氟甲基)苯甲酸 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
α,α,α-三氟对甲苯酸, 对三氟甲基苯甲酸
188460010
188460050
ACRO188460010EA
0
CNY
ACRO188460010
ACRO188460050
4-(三氟甲基)苯甲酸 98%
4-(三氟甲基)苯甲酸
分子式:
C₈H₅F₃O₂ 分子量: 190.12 g/mol 溶点: 219…222 °C |
MDL编号:
MFCD00002562 CAS号: 455-24-3 EINECS: 207-242-8 |
产品技术规格
Appearance | White to light grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 219 to 222 °C |
GC | ≥97.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...