2-(三氟甲基)苯甲酰氯 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
α,α,α-三氟邻甲基苯甲酰氯
312930050
312930010
ACRO312930050EA
0
CNY
ACRO312930050
ACRO312930010
2-(三氟甲基)苯甲酰氯 98%
2-(三氟甲基)苯甲酰氯
分子式:
C₈H₄ClF₃O 分子量: 208.57 g/mol 沸点: 84…85 °C (16 mmHg) 溶点: –22 °C 密度: 1.41 g/cm³ 闪点: 95 °C 存储条件: Ambient |
MDL编号:
MFCD00000667 CAS号: 312-94-7 EINECS: 206-233-6 UN号: 3265 ADR: 8 II |
产品技术规格
Appearance | Clear yellow to pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4780 to 1.4810 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...