苄腈 99+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 苯乙腈

180940010 180945000
ACRO180940010EA 0 CNY
ACRO180940010 ACRO180945000
苄腈 99+%
苄腈
分子式: (C₆H₅)CH₂CN
沸点: 234 °C (1013 hPa)
溶点: –24 °C
密度: 1,01 g/cm³
闪点: 102 °C
存储条件: Ambient
MDL编号: MFCD00001894
CAS号: 140-29-4
EINECS: 205-410-5
UN号: 2470
ADR: 6.1,III
Merck Index: 13,01133

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5240 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR