4-氯-2-甲基苯胺 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-氨基-5-氯甲苯, 4-氯邻甲苯胺

109370050 109371000
ACRO109370050EA 0 CNY
ACRO109370050 ACRO109371000
4-氯-2-甲基苯胺 98%
4-氯-2-甲基苯胺
分子式: C₇H₈ClN
分子量: 141.6 g/mol
沸点: 241
溶点: 29…30 °C
密度: 1.19 g/cm³
闪点: 124 °C
MDL编号: MFCD00007842
CAS号: 95-69-2
EINECS: 202-441-6
UN号: 2239
ADR: 6.1 III

现在订购

产品技术规格

Appearance Clear brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5865 (20°C, 589 nm)
GC ≥97.5 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR