5-Chloro-o-toluidine 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-Chloro-2-methylaniline, 2-Amino-4-chlorotoluene

109380050 109381000 109385000
ACRO109380050EA 0 CNY
ACRO109380050 ACRO109381000 ACRO109385000
5-Chloro-o-toluidine 98%
5-Chloro-o-toluidine
分子式: C₇H₈ClN
分子量: 141.6 g/mol
沸点: 237
溶点: 22
闪点: 160
存储条件: Ambient
MDL编号: MFCD00007779
CAS号: 95-79-4
EINECS: 202-452-6
UN号: 2239
ADR: 6.1 III

现在订购

产品技术规格

Appearance Clear yellow to light brown Liquid after melting (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5850 (20°C, 589 nm)
GC ≥97.5 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR