α-Chloro-3,4-difluorotoulene 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 1-(Chloromethyl)-3,4-difluorobenzene, 3,4-Difluorobenzyl chloride, 4-(Chloromethyl)-1,2-difluorobenzene

384420050 384420250
ACRO384420050EA 0 CNY
ACRO384420050 ACRO384420250
α-Chloro-3,4-difluorotoulene 98%
α-Chloro-3,4-difluorotoulene
分子式: C₇H₅ClF₂
分子量: 162.57 g/mol
MDL编号: MFCD00236218
CAS号: 698-80-6
UN号: 3261
ADR: 8 II

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4940 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR