2-溴苄醇 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(2-Bromophenyl)methanol
180100100
180100250
ACRO180100100EA
0
CNY
ACRO180100100
ACRO180100250
2-溴苄醇 98%
2-溴苄醇
分子式:
C₇H₇BrO 沸点: 255 °C (1013 hPa) 溶点: 78…82 °C |
MDL编号:
MFCD00004600 CAS号: 18982-54-2 EINECS: 242-719-4 |
产品技术规格
Appearance | White to beige Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 78 to 82 °C |
GC | ≥97.5 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...