2-溴-4′-氯苯乙酮 98+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
ω-溴-4-氯苯乙酮, 4-氯苯酰甲基溴
146360250
146361000
ACRO146360250EA
0
CNY
ACRO146360250
ACRO146361000
2-溴-4′-氯苯乙酮 98+%
2-溴-4′-氯苯乙酮
分子式:
C₈H₆BrClO 分子量: 233.49 g/mol 沸点: 278 溶点: 95…99 °C |
MDL编号:
MFCD00000625 CAS号: 536-38-9 EINECS: 208-631-5 UN号: 3261 ADR: 8 II Merck Index: 13,02172 |
产品技术规格
Appearance | White to light yellow Crystals |
Infrared spectrum | Conforms |
Melting point | 95 to 99 °C |
Loss on drying | ≤1 % (70 °C, 2 h) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...