4-甲酰基苯硼酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 4-醛基苯硼酸, 对甲酰苯硼酸

359380010 359380050
ACRO359380010EA 0 CNY
ACRO359380010 ACRO359380050
4-甲酰基苯硼酸 97%
4-甲酰基苯硼酸
分子式: C₇H₇BO₃
溶点: 256…257 °C
存储条件: Refrigerator
MDL编号: MFCD00151823
CAS号: 87199-17-5

现在订购

产品技术规格

Appearance White to yellow to beige Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR