1-氯-4-乙炔基苯 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(4-氯苯基)乙炔
377060010
377062500
ACRO377060010EA
0
CNY
ACRO377060010
ACRO377062500
1-氯-4-乙炔基苯 98%
1-氯-4-乙炔基苯
分子式:
C₈H₅Cl 沸点: 79…82 °C (23 mmHg) 溶点: 43…47 °C 闪点: 10 °C |
MDL编号:
MFCD00191917 CAS号: 873-73-4 UN号: 1325 ADR: 4.1,II |
产品技术规格
Appearance | Yellow to light brown Crystalline mass |
Infrared spectrum | Conforms |
Melting point | 43 to 47 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...