反式-1,2-二苯乙烯 96%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 反-1,2-二苯乙烯

161041000 161045000
ACRO161041000EA 0 CNY
ACRO161041000 ACRO161045000
反式-1,2-二苯乙烯 96%
反式-1,2-二苯乙烯
分子式: C₁₄H₁₂
沸点: 305…307 °C (744 mmHg)
溶点: 122…126 °C
密度: 0,9707 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00064300
CAS号: 103-30-0
EINECS: 203-098-5
UN号: 3077
ADR: 9,III
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 122 to 126 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR