甲基苯基硫醚 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 甲基苯基硫醚, 茴香硫醚

138670250 138671000 138675000
ACRO138670250EA 0 CNY
ACRO138670250 ACRO138671000 ACRO138675000
甲基苯基硫醚 99%
甲基苯基硫醚
分子式: C₇H₈S
分子量: 124.21 g/mol
沸点: 188 °C (760 mmHg)
溶点: –15 °C
密度: 1.05 g/cm³
闪点: 73 °C
存储条件: Ambient
MDL编号: MFCD00008559
CAS号: 100-68-5
EINECS: 202-878-2
UN号: 3082
ADR: 9 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5880 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -8° to -12° (20°C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR