二苯醚 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
苯醚, 联苯醚
130600010
130605000
ACRO130600010EA
0
CNY
ACRO130600010
ACRO130605000
二苯醚 99%
二苯醚
分子式:
C₁₂H₁₀O 分子量: 170.21 g/mol 沸点: 259 °C (760 mmHg) 溶点: 26…30 °C 密度: 1.073 g/cm³ 闪点: 115 °C 存储条件: Ambient |
MDL编号:
MFCD00003034 CAS号: 101-84-8 EINECS: 202-981-2 UN号: 3077 ADR: 9 III Merck Index: 12,07442 |
产品技术规格
Appearance | Clear light yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5795 to 1.5815 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...