3,4-二甲氧基苯磺酰氯 97%

供应商: Thermo Fisher Scientific
Danger

354390010 354390050
ACRO354390010EA 0 CNY
ACRO354390010 ACRO354390050
3,4-二甲氧基苯磺酰氯 97%
3,4-二甲氧基苯磺酰氯
分子式: C₈H₉ClO₄S
溶点: 65…70 °C
存储条件: Ambient
MDL编号: MFCD00051769
CAS号: 23095-31-0
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 65 to 70 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR