乙氧基苯 98+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
苯乙醚, 乙基苯基醚, 乙氧基苯
221491000
221492500
221495000
ACRO221491000EA
0
CNY
ACRO221491000
ACRO221492500
ACRO221495000
乙氧基苯 98+%
乙氧基苯
分子式:
C₆H₅OC₂H₅ 沸点: 169…170 °C (760 mmHg) 溶点: –30 °C 密度: 0,96 g/cm³ 闪点: 57 °C 存储条件: Ambient |
MDL编号:
MFCD00009090 CAS号: 103-73-1 EINECS: 203-139-7 UN号: 3271 ADR: 3,III Merck Index: 12,07374 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5055 to 1.5085 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...