2-氨基苯硫酚 98%
供应商: Thermo Fisher Scientific
|
Danger
|
153901000
153905000
ACRO153901000EA
0
CNY
ACRO153901000
ACRO153905000
2-氨基苯硫酚 98%
2-氨基苯硫酚
分子式:
C₆H₇NS 沸点: 234 °C (1013 hPa) 溶点: 20 °C 密度: 1,16 g/cm³ 闪点: 79 °C 存储条件: Refrigerator |
MDL编号:
MFCD00007702 CAS号: 137-07-5 EINECS: 205-277-3 UN号: 3267 ADR: 8,II |
产品技术规格
Appearance | Clear yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6410 to 1.6450 (20°C, 589 nm) |
Titration Argentometric | 97.5 to 102.5 % |
Additional info | oxidized (disulphide) compound is steadily formed in air |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...