4-氨基苯酚 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 对羟基苯胺

104270010 104270050 104272500
ACRO104270010EA 0 CNY
ACRO104270010 ACRO104270050 ACRO104272500
4-氨基苯酚 97%
4-氨基苯酚
分子式: H₂NC₆H₄OH
分子量: 109.13 g/mol
沸点: 284 °C (760 mmHg)
溶点: 187…191 °C
闪点: 189 °C
存储条件: Ambient
MDL编号: MFCD00007869
CAS号: 123-30-8
EINECS: 204-616-2
UN号: 2512
ADR: 6.1 III
Merck Index: 13,00461
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7af17b754d4ebaa0993b4544d4e6ab6c116724c&vid=6275667c2624ee686f61715acd420dc596e076fa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to cream to beige to light brown Powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR