4-(三氟甲氧基)苯胺 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
α,α,α-三氟对茴香胺
297640025
297640250
ACRO297640025EA
0
CNY
ACRO297640025
ACRO297640250
4-(三氟甲氧基)苯胺 99%
4-(三氟甲氧基)苯胺
分子式:
C₇H₆F₃NO 分子量: 177.13 g/mol 沸点: 83…85 °C (15 mmHg) 密度: 1.31 g/cm³ 闪点: 80 °C |
MDL编号:
MFCD00041314 CAS号: 461-82-5 EINECS: 207-317-5 UN号: 2810 ADR: 6.1 II |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4620 to 1.4640 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...