2,3,4,5,6-五氟苯甲酰氯 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 五氟苯甲酰氯, 五氟苯酰氯

147120050 147120250
ACRO147120050EA 0 CNY
ACRO147120050 ACRO147120250
2,3,4,5,6-五氟苯甲酰氯 98%
2,3,4,5,6-五氟苯甲酰氯
分子式: C₇ClF₅O
沸点: 158 °C (1013 hPa)
密度: 1,6 g/cm³
存储条件: Ambient
MDL编号: MFCD00000657
CAS号: 2251-50-5
EINECS: 218-842-4
UN号: 3265
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
Titration with HCl 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR