2,4-二氟苯硼酸 98%
供应商: Thermo Fisher Scientific
|
Warning
|
359130010
359130100
ACRO359130010EA
0
CNY
ACRO359130010
ACRO359130100
2,4-二氟苯硼酸 98%
2,4-二氟苯硼酸
分子式:
C₆H₅BF₂O₂ 溶点: 247…250 °C 存储条件: Refrigerator |
MDL编号:
MFCD01318998 CAS号: 144025-03-6 |
产品技术规格
Appearance | White to yellow to light beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 247 to 251 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...