2-氨基苯甲酸乙酯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
邻氨基苯甲酸乙酯
222231000
222235000
ACRO222231000EA
0
CNY
ACRO222231000
ACRO222235000
2-氨基苯甲酸乙酯 99%
2-氨基苯甲酸乙酯
分子式:
C₉H₁₁NO₂ 沸点: 129…130 °C (9 mmHg) 溶点: 13…15 °C 密度: 1,117 g/cm³ 闪点: >110 °C |
MDL编号:
MFCD00007711 CAS号: 87-25-2 EINECS: 201-735-1 |
产品技术规格
Appearance | Clear colorless to yellow to amber Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5630 to 1.5690 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...