水杨酰肼 98+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 水杨酸酰肼, 2-羟基苯甲酰肼

132610250 132611000
ACRO132610250EA 0 CNY
ACRO132610250 ACRO132611000
水杨酰肼 98+%
水杨酰肼
分子式: C₇H₈N₂O₂
分子量: 152.15 g/mol
溶点: 147…150 °C
MDL编号: MFCD00007599
CAS号: 936-02-7
EINECS: 213-311-3

现在订购

产品技术规格

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 151 °C
Titration with HClO4 ≥98.0 %
Specific optical rotation -14° to -18° (20 °C, 589 nm) (c=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR