Anthranilamide, Sigma-Aldrich®

供应商: Merck
Warning

A89804-100G A89804-5G
SIALA89804-100GEA 0 CNY
SIALA89804-100G SIALA89804-5G
Anthranilamide, Sigma-Aldrich®
2-氨基苯甲酰胺
分子式: C₇H₈N₂O
沸点: 300 °C (1013 hPa)
溶点: 105…107 °C
闪点: >185 °C
存储条件: Ambient
MDL编号: MFCD00007981
CAS号: 88-68-6
EINECS: 201-851-2

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR