2-氨基苯甲酰胺 99+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 邻氨基苯甲酰胺, 2-氨基苯甲酰胺, 2-AB

104900050 104901000
ACRO104900050EA 0 CNY
ACRO104900050 ACRO104901000
2-氨基苯甲酰胺 99+%
2-氨基苯甲酰胺
分子式: C₇H₈N₂O
分子量: 136.15 g/mol
沸点: 300 °C (760 mmHg)
溶点: 111…114 °C (dec.)
闪点: 198 °C
存储条件: Ambient
MDL编号: MFCD00007981
CAS号: 88-68-6
EINECS: 201-851-2

现在订购

产品技术规格

Appearance Light brown to grey-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 111 to 114 °C
HPLC ≥99.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR