苯甲酰溴 97%
供应商: Thermo Fisher Scientific
|
Danger
|
160950050
160950250
ACRO160950050EA
0
CNY
ACRO160950050
ACRO160950250
苯甲酰溴 97%
苯甲酰溴
分子式:
C₇H₅BrO 沸点: 218…219 °C (760 mmHg) 溶点: –24 °C 密度: 1,57 g/cm³ 闪点: 90 °C 存储条件: Ambient |
MDL编号:
MFCD00000112 CAS号: 618-32-6 EINECS: 210-544-2 UN号: 3265 ADR: 8,II |
产品技术规格
Appearance | Clear colorless to yellow to light orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5880 to 1.5920 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...