4-氯苯基硼酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 4-氯苯硼酸

344410010 344410050
ACRO344410010EA 0 CNY
ACRO344410010 ACRO344410050
4-氯苯基硼酸 97%
4-氯苯基硼酸
分子式: C₆H₆BClO₂
溶点: 259…262 °C
密度: ~ 1 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00039137
CAS号: 1679-18-1
EINECS: 216-845-5

现在订购

产品技术规格

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR