2,3-二氯苯硼酸 97%
供应商: Thermo Fisher Scientific
|
Warning
|
367710010
367710050
367710250
ACRO367710010EA
0
CNY
ACRO367710010
ACRO367710050
ACRO367710250
2,3-二氯苯硼酸 97%
2,3-二氯苯硼酸
分子式:
C₆H₅BCl₂O₂ 分子量: 190.82 g/mol 溶点: 242…244 °C 存储条件: Ambient |
MDL编号:
MFCD01075703 CAS号: 151169-74-3 |
产品技术规格
Appearance | White to cream Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...