4-溴-N,N-二甲基苯胺 99%

供应商: Thermo Fisher Scientific
Warning

222210250 222211000
ACRO222210250EA 0 CNY
ACRO222210250 ACRO222211000
4-溴-N,N-二甲基苯胺 99%
4-溴-N,N-二甲基苯胺
分子式: C₈H₁₀BrN
分子量: 200.08 g/mol
沸点: 264
溶点: 53…56 °C
闪点: >112 °C
存储条件: Ambient
MDL编号: MFCD00000093
CAS号: 586-77-6
EINECS: 209-582-2
UN号: 2811
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to grey to light blue Crystalline solid
Infrared spectrum Conforms
Melting point 53 to 56 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR