2-甲氧基苯基硼酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 邻甲氧基苯硼酸

344660010 344660050
ACRO344660010EA 0 CNY
ACRO344660010 ACRO344660050
2-甲氧基苯基硼酸 97%
2-甲氧基苯基硼酸
分子式: C₇H₉BO₃
分子量: 151.96 g/mol
溶点: 106…111 °C
存储条件: Refrigerator
MDL编号: MFCD00236047
CAS号: 5720-06-9

现在订购

产品技术规格

Appearance Off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 106 to 111 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR