吲哚啉 98%
供应商: Thermo Fisher Scientific
|
Warning
|
122240250
122241000
ACRO122240250EA
0
CNY
ACRO122240250
ACRO122241000
吲哚啉 98%
吲哚啉
分子式:
C₈H₉N 沸点: 220…221 °C (760 mmHg) 溶点: –21 °C 密度: 1,063 g/cm³ 闪点: 92 °C 存储条件: Ambient |
MDL编号:
MFCD00005705 CAS号: 496-15-1 EINECS: 207-816-8 |
产品技术规格
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5910 to 1.5950 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...