7-氟靛红 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
7-氟-2,3-吲哚二酮
F0551-25G
F0551-5G
TCIAF0551-25GEA
0
CNY
TCIAF0551-25G
TCIAF0551-5G
7-氟靛红 ≥98.0% (by GC, titration analysis)
7-氟靛红
分子式:
C₈H₄FNO₂ 分子量: 165.12 g/mol |
MDL编号:
MFCD01569508 CAS号: 317-20-4 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 193.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...